Somso® Nursing Care Manikins
470233-238
- Descripción:Advanced Nursing Care Manikin
- Longitud:178 cm
- Referencia:470233-238
- Peso:23.5 kg
For more advanced patient care practice!
- A great starter manikin
- Expandable over time
Standard nursing doll in natural size for nursing care comprising of the following parts: Removable head, removable eyes, dentures and tracheostomy, thorax with chest wall and moving arms with infusion and injection pads, lower body with abdominal wall and anus praeter abdominal wall, injection pads in the buttocks, movable mounted legs with injection pads in the thigh, lung alveolus, stomach, intestines, detachable female and male genitalia with rectum and bladder.
The advanced version includes lungs exhibiting closure, additional male genetalia and a higher capacity bladder (2 L).