Orders LinkContactUsLinkComponent
Corning® Pipette Tips, Microvolume
 :   Corning
Corning® Pipette Tips, Microvolume
 :  Corning
Corning® Pipette Tips, Microvolume
Corning® Pipette Tips, Microvolume
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
  • Catalog No:
  • 29443-298
  • 29443-304
  • 29443-320
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Product Family Options
  • Volume
    Sterile
    Tip Color
    Packed
  • 0.1 - 10 µl
    -
    Natural
    Rack Packed
    Case (960UN)
    Case (960UN) - $1,540.00
    $1,540.00
    Case (960UN)
       $1,540.00
     : 4826
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    29443-298
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 0.2 - 10 µl
    -
    Natural
    Bulk Packed
    Case (10,000UN)
    Case (10,000UN) - $9,720.00
    $9,720.00
    Case (10,000UN)
       $9,720.00
     : 4840
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    29443-304
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 0.1 - 10 µl
    +
    Natural
    Rack Packed
    Case (960UN)
    Case (960UN) - $2,250.00
    $2,250.00
    Case (960UN)
       $2,250.00
     : 4894
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    29443-320
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact