Orders LinkContactUsLinkComponent
Carboy Replacement Spigot, Polypropylene, Dynalon
Carboy Replacement Spigot, Polypropylene, Dynalon
Carboy Replacement Spigot, 60.3×38.1×85.7 mm
Catalog # 490007-212
Supplier:   DYNALON
Número de proveedor #:  505634-7
Carboy Replacement Spigot, Polypropylene, Dynalon
Catalog # 490007-212
Supplier:  DYNALON
Carboy Replacement Spigot, Polypropylene, Dynalon
Carboy Replacement Spigot, Polypropylene, Dynalon
Carboy Replacement Spigot, 60.3×38.1×85.7 mm
Price
$1,167.46
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Replacement autoclavable, polypropylene spigot for the Azlon carboy series.
  • Spigot features removable tube fitting for added convenience
  • Durable and leak-proof, heavy-duty spigot with boss fitting for extra strength
This polypropylene spigot consists of two parts, an ON/OFF tab with screw, and a housing with integrated silicone gasket. The housing screws onto the boss of the carboy. Engage the thread a few turns and then push down on the spigot to seat the gasket over the boss. Then tighten the threads while continuing to apply pressure down on the spigot. This should result in a tight and leak-proof fit.
For cleaning and autoclaving the polypropylene spigot, the ON/OFF tab will unscrew from the housing. In the ON position, just lift and push the tab counterclockwise up and over the white stop on the housing. It will unscrew completely when turned 180°. Reverse the process for reassembly. Cleaning is important as sediments may eventually build up. Autoclave as needed.
Especificaciones
  • Description:
    Carboy Replacement Spigot
  • L×W×H:
    60.3×38.1×85.7 mm