Orders LinkContactUsLinkComponent
Erlenmeyer Flask, Polypropylene, Dynalon
250 ml, 25 ml, 19/26
  30621-006
 :   DYNALON
 :  241345
Erlenmeyer Flask, Polypropylene, Dynalon
  30621-006
 :  DYNALON
Erlenmeyer Flask, Polypropylene, Dynalon
Erlenmeyer Flask, Polypropylene, Dynalon
250 ml, 25 ml, 19/26
$2,180.49
Case (6UN)
Autoclavable polypropylene Erlenmeyer flask with molded graduations offers excellent chemical resistance.
Graduations are permanently molded into the flask.
  • Graduation Interval:
    25 ml
  • ST Joint No.:
    19/26
  • Capacity:
    250 ml