Avantor® Genesis, Cyano HPLC Columns
Catalog # 75799-006
Supplier: Avantor
Specifications
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:4 µm
- Longitud:150 mm
- Extremo tapado:Yes
- Código USP:L18
- Rango de pH:1 -10
- Surface area:300 m²/g
- Carga de carbono:7%
- Column Hardware:Acero inoxidable
- Referencia:75799-006
- Packing Material:Silica
- Phase:Cyano
- Nº de proveedor:FM15965E
Specifications
About this item
Avantor® Genesis phases are based on high purity metal-free spherical silica.
- Good peak shape and reproducibility
- Long column lifetime
- pH stability 1 to 10
These are suitable for the analysis of a wide range of analytes.