Storage Vials, Microsolv™
Autosampler Vials, 20.00 ml, 57×27.5 mm, Borosilicate glass, Amber, No, 24-400
Catalog # 97058-114
Supplier: MICROSOLV TECH CORP
Número de proveedor #: 97020-0AV
Jump to:
Product Details & Documents
Screw top vials made up of high quality glass for long term storage.
- Can be used for EPA testing
- Vials and caps are used to store chemical compounds for years
Specifications
- Cap included:No
- Cap Size:24-400
- Capacity:20.00 ml
- Color:Amber
- Description:Autosampler Vials
- Material:Borosilicate glass
- Diameter ×H:57×27.5 mm