Avantor® Genesis, C8 HPLC Columns
Analytical, C8, 5 µm, 4.60 mm, 250 mm, Silica
Catalog # 76398-924
Supplier: Avantor
Supplier #: GEN-5C8-250D
Jump to:
Product Details & Documents
Avantor® Genesis phases are based on high purity metal-free spherical silica.
- Good peak shape and reproducibility
- Long column lifetime
- pH stability 1 to 10
- Stainless steel column hardware
They are suitable for the analysis of a wide range of analytes.
Especificaciones
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Length:250 mm
- Packing Material:Silica
- Phase:C8