Vydac 214TP C4, HPLC Columns
: Avantor
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L26
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
- Catalog No:
- 75798-766
- 75798-756
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Código USP:
- L26
- L26
- Certificación ambiental:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)