Teacher Inorganic and Organic Molecular Model Set, 101 Pieces
Catalog # 470231-424
Supplier: EISCO SCIENTIFIC LLC
Specifications
- Descripción:Teacher Molecular Model Set, 121 Pieces
- Referencia:470231-424
Specifications
About this item
Perfect Set for Introductory Chemistry Classroom
- 51 atoms
- 70 bonds
Contents: 10 Black Carbon (4 Hole, 22 mm), 15 White Hydrogen (1 Hole, 15 mm), 6 Red Oxygen (2 Hole, 20 mm), 4 Blue Nitrogen (4 Hole, 22mm), 4 Green Chlorine (1 Hole, 20 mm), 1 Green Chlorine (4 Hole, 22 mm), 1 Yellow Sulfur (2 Hole, 20 mm), 1 Yellow Sulfur (4 Hole, 22 mm), 1 Purple Phosphorus (4 Hole, 22 mm), 2 Orange Bromine (1 Hole, 20 mm), 2 Purple Iodine (1 Hole, 20 mm), 2 Grey Metal (1 Hole, 15 mm), 1 Grey Metal (2 Hole, 20 mm), 1 Grey Metal (4 Hole, 22 mm), 10 Grey Long Bonds, 30 Grey Medium Bonds, 30 Grey Short Bonds.