Basic Inorganic and Organic Chemistry Molecular Model Set, 78 Pieces
Catálogo # 470231-422
Proveedor: EISCO SCIENTIFIC LLC
Especificaciones
- Descripción:Basic Molecular Model Set, 78 Pieces
- Referencia:470231-422
Especificaciones
Acerca de este artículo
Standard Size Molecular Model Set
- 54 atoms
- 24 links
Contents: 24 White Hydrogen (1 Hole, 15 mm), 12 Black Carbon (4 Hole, 22 mm), 6 Red Oxygen (2 Hole, 20 mm), 2 Blue Nitrogen (4 Hole, 22 mm), 4 Blue Nitrogen (3 Hole, 22 mm), 1 Yellow Sulfur (4 Hole, 22 mm), 1 Yellow Sulfur (2 Hole, 22 mm), 2 Green Chlorine (1 Hole, 22 mm), 1 Brown Bromine (1 Hole, 22 mm), 1 Purple Iodine (1 Hole, 22 mm), 16 Medium Grey Bonds, 8 Long Grey Bonds.