Especificaciones
- Descripción:Molecular Model Set, 253 Atoms
- Referencia:470231-440
Especificaciones
Acerca de este artículo
Tutorial Universal Set
- 259 atoms
- 180 links
- 3 orbitals
Larger set allows for a large number of organic, inorganic and polymer (polystyrene, PVC, PP) molecules to be made simultaneously.
Contents: 96 White Hydrogen (Single Bonded, 15 mm), 50 Black Carbon (Triple Bonded, 20 mm), 18 Dark Grey Carbon (Double Bonded, 20 mm), 4 Light Grey Carbon (Triple Bonded, 20 mm), 2 Blue Nitrogen (Triple Bonded, 20 mm), 2 Blue Nitrogen (Triple Bonded, 20 mm), 4 Blue Nitrogen (Single Bonded, 20 mm), 4 Blue Nitrogen (Triple Bonded, 20 mm), 8 Blue Nitrogen (Single Bonded, 20 mm), 8 Blue Nitrogen (Triple Bonded, 20 mm), 24 Red Oxygen (Single Bonded Bent, 20 mm), 1 Red Oxygen (Triple Bonded, 20 mm), 6 Red Oxygen (Double Bonded, 20mm), 4 Red Oxygen (Double Bonded, 20 mm), 6 Yellow Sulfur (Triple Bonded, 20 mm), 4 Yellow Sulfur (Single Bonded Bent, 20 mm), 2 Green Halogen (Triple Bonded, 20 mm), 10 Green Halogen (Single Bonded, 20 mm), 120 Simple Links (52 mm), 36 Dual Links (52 mm), 18 Grey Turning Links (72 mm), 6 White H-Link (bridge), 2 Red Orbitals (46 mm), 1 Blue Orbital (46 mm).