Orders LinkContactUsLinkComponent
VWR® Digital Handheld Refractometers
0 - 95 °Bx , 1,33 - 1,54 nD
Catalog # 75997-574
Supplier:   VWR International
VWR® Digital Handheld Refractometers
Catalog # 75997-574
Supplier:  VWR International
VWR® Digital Handheld Refractometers
VWR® Digital Handheld Refractometers
0 - 95 °Bx , 1,33 - 1,54 nD
Price
$19,601.28
Log in for availability
Each (1UN)
Product Details & Documents
Dual scale handheld refractometers designed to provide ranges of Brix (sugar concentration) and refractive index measurements.
  • Optical glass prism and stainless steel dish to stabilize sample temperature
  • Simple zero calibration with distilled water
  • Automatic temperature compensation (ATC) to correct readings of water and sucrose solutions to 20°C
 : Supplied with two batteries and a battery indicator, multi-language guide and protective case.
Specifications
  • Range:
    0 - 95 °Bx , 1,33 - 1,54 nD
  • Cat. No.:
    75997-574
  • Accuracy:
    ±0,0003 nD ; ±0,2 °Brix
  • W×D×H:
    55 × 120 × 35 mm
  • Measurement time:
    2 seconds
  • Weight:
    85 g
  • Resolution:
    0,1 °Bx ; 0,0001 nD