Orders LinkContactUsLinkComponent
Difco™ M17 Agar, BD Diagnostics
M 17 Agar, 500 g
  95056-558
Difco™ M17 Agar, BD Diagnostics
  95056-558
 :  
Difco™ M17 Agar, BD Diagnostics
M 17 Agar, 500 g
 :  
$14,600.00
Each (1UN)
Use for the cultivation and isolation of mesophilic lactic streptococci and for the selection of Streptococcus thermophilus from yogurt, cheese starters and other dairy products.
  • Description:
    M 17 Agar
  • Size:
    500 g