Ward's® Essentials Trebuchet Kit
Ward's® Essentials Trebuchet Kit, 1
Catalog # 470229-062
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: VWRGP00009
Jump to:
Product Details & Documents
Recreate the medieval weapon using counterweights.
- Durable materials for years of use
- Build problem-solving, inquiry, and engineering skills
- Aligned with NGSS MS-PS2-2
Students will build a trebuchet to determine the kinetic energy of a projectile. Material: High quality precision-cut Birch verner pure bond plywood, Designed to be safe and robust, Dry firings and heavy projectiles are no problem, Holes are pre drilled, Medieval weapons, engineering, and physics. 21" beam arm.
Especificaciones
- Description:Ward's® Essentials Trebuchet Kit
- No. of Lab Groups:1