PTFE Scoop, Bürkle
PTFE Scoop, 10 ml (0.34 oz.), 34 mm (1.34"), 25 mm (0.98")
Catalog # 76049-272
Supplier: BURKLE INC
Número de proveedor #: 5389-0010
Jump to:
- El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
- Return Policy
Product Details & Documents
All parts coming into contact with the sample medium are made of pure white PTFE
The rod is stabilized with a steel core that is completely encapsulated in PTFE. The 60 cm (23.62") long rod can be extended to 120 cm (47.24") by adding the extension component. The 10 mL (0.34 oz.) PTFE scoop has a strong rod with a length of 155 mm (6.10").
Completely inert against all substances (with the exception of elementary fluoride).
Documents
Certificates
Buscar por número de loteEspecificaciones
- Ø:25 mm (0.98")
- Capacity:10 ml (0.34 oz.)
- Description:PTFE Scoop
- Length:34 mm (1.34")