Friction Cube with Four Surfaces
Friction Cube with Four Surfaces
Catalog # 470230-400
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH1120151
Jump to:
Product Details & Documents
Ideal for exploring force of friction.
- 2 inch cube with hook
- Surfaces: Wood, Paper, Vinyl, Sandpaper
- By pulling the cube by the attached hook, one can both qualitatively and quantitatively demonstrate the properties of static and kinetic friction
The friction cube provides four surfaces for exploration into the force of friction. Each side of the 2" cube provides a surface (wood, paper, vinyl, sandpaper) with a different coefficient of friction. By pulling the cube by the attached hook, one can both qualitatively and quantitatively demonstrate the properties of static and kinetic friction.
Especificaciones
- Description:Friction Cube with Four Surfaces