Orders LinkContactUsLinkComponent
VWR® Basic Industrial Lab Coats
VWR® Basic Industrial Lab Coats
Supplier:  VWR International
Certificates
VWR® Basic Industrial Lab Coats
35 Options Available Below
Product Details & Documents
Disposable lab coat made from 20 gram spunbond polypropylene (SPP) fabric. Provides lightweight, breathable protection for non-hazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.
  • Lightweight and breathable fabric reduces heat buildup during extended use
    Snap front closure for easy donning and removal
  • Elastic wrists and ankles for better coverage
  • Single-use, disposable design for contamination control
  • Economical solution for basic protection needs
  • Non-linting material helps maintain clean environments
  • Suitable for general industrial, food processing, and non-hazardous environments
The VWR® basic protection lab coat is designed to provide reliable, economical protection in environments where contamination control is essential but exposure risks are minimal. Constructed from 20 gram spunbond polypropylene (SPP), this garment offers a lightweight and breathable solution that ensures user comfort during extended wear. The non-woven SPP fabric is non-linting, reducing the risk of particle contamination, and is suitable for clean areas, food processing, and general industrial applications.
Key design features include a snap front closure for easy donning and removal, and an elastic cuff option is also available. The disposable nature of the garment supports hygiene compliance.
 : Lab coats are packaged 30 per case.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76008-420
  • 76204-788
  • 76008-422
  • 76204-790
  • 76008-426
  • 76204-792
  • 76008-428
  • 76204-632
  • 76008-430
  • 76204-634
  • 76008-432
  • 76204-636
  • 76008-434
  • 76204-638
  • 76008-436
  • 76008-438
  • 76008-440
  • 76008-442
  • 76008-480
  • 76008-482
  • 76008-484
  • 76008-486
  • 76008-488
  • 76008-490
  • 76008-492
  • 76008-494
  • 76008-496
  • 76008-498
  • 76008-500
  • 76008-502
  • 76008-504
  • 76008-506
  • 76008-508
  • 76008-510
  • 76008-512
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • Weight:
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
Especificaciones
Product Family Options
Standard lab coats
Información del producto
  • Color
    Clothing Size
    Length
    Chest Size
    Sleeve
    Availability
    Price
  • White
    S
    33½"
    24½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $401.34
    $401.34
    Case (30UN)
       $401.34
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-420
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    M
    35½"
    25½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $410.95
    $410.95
    Case (30UN)
       $410.95
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-422
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    L
    37½"
    26"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $419.48
    $419.48
    Case (30UN)
       $419.48
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-426
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XL
    40"
    26¾"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $422.14
    $422.14
    Case (30UN)
       $422.14
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-428
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XXL
    43¼"
    27½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $428.00
    $428.00
    Case (30UN)
       $428.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-430
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    3XL
    45½"
    28½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $439.73
    $439.73
    Case (30UN)
       $439.73
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-432
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    4XL
    47¼"
    29"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $444.55
    $444.55
    Case (30UN)
       $444.55
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-434
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    S
    33½"
    24½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $415.20
    $415.20
    Case (30UN)
       $415.20
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-436
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    M
    35½"
    25½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $423.75
    $423.75
    Case (30UN)
       $423.75
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-438
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    L
    37½"
    26"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $433.34
    $433.34
    Case (30UN)
       $433.34
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-440
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    XL
    40"
    26¾"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $437.61
    $437.61
    Case (30UN)
       $437.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-442
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    XXL
    43¼"
    27½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $444.55
    $444.55
    Case (30UN)
       $444.55
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-480
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    3XL
    45½"
    28½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $457.32
    $457.32
    Case (30UN)
       $457.32
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-482
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Blue
    4XL
    47¼"
    29"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $462.12
    $462.12
    Case (30UN)
       $462.12
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-484
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    S
    33½"
    24½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $415.20
    $415.20
    Case (30UN)
       $415.20
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-486
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    M
    35½"
    25½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $423.75
    $423.75
    Case (30UN)
       $423.75
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-488
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    L
    37½"
    26"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $433.34
    $433.34
    Case (30UN)
       $433.34
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-490
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    XL
    40"
    26¾"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $437.61
    $437.61
    Case (30UN)
       $437.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-492
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    XXL
    43¼"
    27½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $444.55
    $444.55
    Case (30UN)
       $444.55
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-494
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    3XL
    45½"
    28½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $457.32
    $457.32
    Case (30UN)
       $457.32
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-496
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    4XL
    47¼"
    29"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $462.12
    $462.12
    Case (30UN)
       $462.12
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-498
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    S
    33½"
    24½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $408.27
    $408.27
    Case (30UN)
       $408.27
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-500
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    M
    35½"
    25½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $417.36
    $417.36
    Case (30UN)
       $417.36
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-502
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    L
    37½"
    26"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $426.41
    $426.41
    Case (30UN)
       $426.41
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-504
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    XL
    40"
    26¾"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $430.68
    $430.68
    Case (30UN)
       $430.68
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-506
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    XXL
    43¼"
    27½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $437.61
    $437.61
    Case (30UN)
       $437.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-508
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    3XL
    45½"
    28½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $448.80
    $448.80
    Case (30UN)
       $448.80
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-510
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    4XL
    47¼"
    29"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $453.07
    $453.07
    Case (30UN)
       $453.07
    Especificaciones:
    More
    Specifications
    Cat. No.
    76008-512
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
Snap front with elastic cuffs
Información del producto
  • Color
    Clothing Size
    Length
    Chest Size
    Sleeve
    Availability
    Price
  • White
    S
    33½"
    24½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $417.36
    $417.36
    Case (30UN)
       $417.36
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-788
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    M
    35½"
    25½"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $426.41
    $426.41
    Case (30UN)
       $426.41
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-790
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    L
    37½"
    26"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $435.46
    $435.46
    Case (30UN)
       $435.46
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-792
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XL
    40"
    26¾"
    22"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $437.61
    $437.61
    Case (30UN)
       $437.61
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-632
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XXL
    43¼"
    27½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $444.55
    $444.55
    Case (30UN)
       $444.55
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-634
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    3XL
    45½"
    28½"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $455.73
    $455.73
    Case (30UN)
       $455.73
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-636
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    4XL
    47¼"
    29"
    23"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (30UN)
    Case (30UN) - $460.00
    $460.00
    Case (30UN)
       $460.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    76204-638
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²