Force Table, Economy
Force Table, Economy
Catalog # 470230-142
Supplier: EISCO SCIENTIFIC LLC
Supplier #: PH0347FT
Jump to:
Product Details & Documents
Force table is inexpensive and light in weight.
- Three-legged
- Top is made from durable PVC
Laminated top allows use of erasable markers. The legs screw in the table quite easily and have built-in levelling screws for levelling adjustments.
: Weights not included.
: The table is supplied with 4 special ball bearing pulleys and 4 weight hangers.
Specifications
- Description:Force Table, Economy