Force Table Pulley
Force table pulley
Catalog # 470230-108
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH0300C
Jump to:
Product Details & Documents
Plastic pulley with table clamp, includes hand knob for tightening.
- Plastic 2" pulley
- Clamp made of hard plastic
- Clamp max opening 1.250"
- Grey hand knob for tightening clamp
labs low friction force table 2" pulley - Clamp max opening is 1.25" Clamp is made of hard plastic. Grey hand knob for tighten clamp
Ordering information: The corresponding force table (470014-356) is also available for purchase.
Especificaciones
- Description:Force table pulley