Pulley with Adjustable Clamp
Pulley with AdjusTable Clamp
Catalog # 470230-104
Supplier: EISCO SCIENTIFIC LLC
Supplier #: PH0297A
Jump to:
Product Details & Documents
Metal pulley with rotatable pulley for experiment set up.
- Sturdy metal construction throughout
- Ideal for demonstration of wide variety of mechanics principles
- 2" (5 cm) dia. sheave
- Completely rotatable pulley for flexibility in experiment setups
- Adjustable to 9" height with 1.9" maximum clamp opening
Sturdy, metal clamp pulley can be used to demonstrate a wide variety of mechanics principles. The metal sheave is 5 cm in diameter and can be adjusted from flush with the tabletop to 13 cm above it. Pulley can be rotated to 360º to suit any experimental setup.
Specifications
- Description:Pulley with AdjusTable Clamp