Ward’s® Automatic Autoclave Series
Acerca de este artículo
Three convenient options for benchtop sterilization.
- Rapid, complete sterilization for educational/research use
- Built-in water tank doesn't require external water supply
- Safety interlocks prevent door from opening while pressurized
- Stainless steel sterilization chambers
- One-year warranty
Ward's® BioClaves offer three preset standard cycle options, for sterilization of liquids, wrapped instruments or unwrapped instruments/plastic ware. A fourth option "Dry Only" is also available for additional drying time to be added to the end of a cycle.
Ward’s® BioClave Mini™: The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. Temperature can be selected at either 121°C or 134°C, and the sterilization time is fully adjustable.
Ward’s® BioClave 18™: The 18 liter BioClave offers preset cycle options (standard programs), for use with liquids, wrapped research instruments or unwrapped instruments/plasticware. An additional option “Dry Only” is also available for additional drying time to be added to the end of a cycle. For special applications, requiring variations in cycle parameters, all settings on the BioClave 18 are fully adjustable.
Ward’s® BioClave 28™- Features an increased chamber size with a diameter of 32cm. With this larger internal chamber, the BioClave 28 is our only model capable of sterilizing 1 and 2 liters flasks (and bottles). Despite the increased chamber size, the unit is still relatively compact, suitable for benchtop use.”
Caution: Not intended for medical use or with food.
2 Opciones disponibles a continuación
Product Details & Documents
Three convenient options for benchtop sterilization.
- Rapid, complete sterilization for educational/research use
- Built-in water tank doesn't require external water supply
- Safety interlocks prevent door from opening while pressurized
- Stainless steel sterilization chambers
- One-year warranty
Ward's® BioClaves offer three preset standard cycle options, for sterilization of liquids, wrapped instruments or unwrapped instruments/plastic ware. A fourth option "Dry Only" is also available for additional drying time to be added to the end of a cycle.
Ward’s® BioClave Mini™: The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. Temperature can be selected at either 121°C or 134°C, and the sterilization time is fully adjustable.
Ward’s® BioClave 18™: The 18 liter BioClave offers preset cycle options (standard programs), for use with liquids, wrapped research instruments or unwrapped instruments/plasticware. An additional option “Dry Only” is also available for additional drying time to be added to the end of a cycle. For special applications, requiring variations in cycle parameters, all settings on the BioClave 18 are fully adjustable.
Ward’s® BioClave 28™- Features an increased chamber size with a diameter of 32cm. With this larger internal chamber, the BioClave 28 is our only model capable of sterilizing 1 and 2 liters flasks (and bottles). Despite the increased chamber size, the unit is still relatively compact, suitable for benchtop use.”
Caution: Not intended for medical use or with food.
Documents
Avantor Top Rated Guide FOCUS Industrial Volume 02 Issue 02
Product Literature
Product Family Options
Bioclaves
Información del producto
- DescriptionCapacityElectricalDimensionsInterior DimensionsSubtotalPrice
- BioClave Mini8 L115 V, 50/60 Hz, 950 W20.0×13.5×12.9"6.7×12.5"Iniciar sesión Para disponibilidadEach (1UN)$165,162.99
- BioClave 1818 L115 V, 50/60 Hz, 1600 W22.0×17.5×15.7"9.8×13.77"Iniciar sesión Para disponibilidadEach (1UN)$229,586.00
Autoclave bags
Información del producto
- Catálogo #ColorW×LSubtotalPrice