Orders LinkContactUsLinkComponent
Eisco® Electrodes
Eisco® Electrodes
Eisco® Electrodes
Certificates
Eisco® Electrodes
Eisco® Electrodes
9 Options Available Below
Product Details & Documents
Individual metal electrodes for electrochemical experiments.
  • Each electrode is stamped with its metal type for easy identification
  • Durable and reusable for multiple lab sessions
Eisco electrodes are an essential tool for students exploring electrochemical reactions and principles. Each electrode strip measures approximately 100×19 mm (4×0.75"), is crafted from high-quality metals and clearly stamped for easy identification.These electrodes are perfect for hands-on learning in topics such as oxidation-reduction reactions, electrode potentials, and galvanic cells.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 470230-330
    Aluminum electrode, strip
    Aluminum electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $66.10
    $66.10
    Each (1UN)
       $66.10
    Supplier Number: PH0927A
  • 470230-332
    Brass electrode, strip
    Brass electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2.10
    $2.10
    Clearance
    Each (1UN)
       $2.10
    Supplier Number: PH0927B
  • 470230-334
    Carbon electrode, rod
    Carbon electrode, rod
    100×5 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $82.62
    $82.62
    Each (1UN)
       $82.62
    Supplier Number: PH0927D
  • 470230-336
    Copper electrode, strip
    Copper electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $110.87
    $110.87
    Each (1UN)
       $110.87
    Supplier Number: PH0927E
  • 470230-338
    Lead electrode, strip
    Lead electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $92.74
    $92.74
    Each (1UN)
       $92.74
    Supplier Number: PH0927F
  • 470230-340
    Nickel electrode, strip
    Nickel electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $251.57
    $251.57
    Each (1UN)
       $251.57
    Supplier Number: PH0927G
  • 470230-342
    Tin electrode, strip
    Tin electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $244.64
    $244.64
    Each (1UN)
       $244.64
    Supplier Number: PH0927H
  • 470230-344
    Zinc electrode, strip
    Zinc electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $115.13
    $115.13
    Each (1UN)
       $115.13
    Supplier Number: PH0927I
  • 470230-346
    Iron electrode, strip
    Iron electrode, strip
    100×19 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $50.10
    $50.10
    Each (1UN)
       $50.10
    Supplier Number: PH0927J