Orders LinkContactUsLinkComponent
Micrometer, Eisco Scientific
Micrometer, 0-25 x 0.01 mm, 0 - 25×0.01 mm
  470230-012
 :  PH0086A
Micrometer, Eisco Scientific
  470230-012
Micrometer, Eisco Scientific
Micrometer, Eisco Scientific
Micrometer, 0-25 x 0.01 mm, 0 - 25×0.01 mm
$396.56
Each (1UN)
Made of nickel plated brass, with ratchet top, accurately machined stainless steel rod.
Micrometer Screw Gauge, Nickel Plated Brass - Range 0-25 x 0.01 mm (1) Micrometer. Range 0-25 x 0.01 mm, supplied in case. Accuracy is perfect for a hobbyist or student -- at 10 mm, accuracy is ± .003 mm.
  • Description:
    Micrometer, 0-25 x 0.01 mm
  • Range:
    0 - 25×0.01 mm
  • Cat. No.:
    470230-012
  • Accuracy:
    ±.003 mm (at 10 mm)