Fluid A Prepared Bottled Media, BD Diagnostics
Catalog # 90004-129
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Descripción:Fluid A, 0.1% Peptic digest of animal tissue
- Tamaño:300 mL
- Referencia:90004-129
- Tipo:Septum/Screw Cap
- Nº de proveedor:290652
Specifications
About this item
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Serum bottle holds up to 100 mL fill volume.
Septum/Screw Cap style has a black, phenolic screw cap with septum.