Orders LinkContactUsLinkComponent
VWR® Round Bottom Flask, 3 Necks, Angled
5000 ml, 295 mm, 223 mm, 29/42, 24/40
  76008-648
VWR® Round Bottom Flask, 3 Necks, Angled
  76008-648
VWR® Round Bottom Flask, 3 Necks, Angled
VWR® Round Bottom Flask, 3 Necks, Angled
5000 ml, 295 mm, 223 mm, 29/42, 24/40
$3,367.84
Case (1UN)
Durable round bottom flasks with 3 necks at an angle.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.
  • Ext.Ø:
    223 mm
  • Capacity:
    5000 ml
  • Height:
    295 mm
  • Center Neck [ST] Joint:
    29/42
  • Side Neck ST Joints:
    24/40