Celestron Handheld Digital Microscope Pro
Product Details & Documents
True 5MP sensor is ideal to capture and save high-resolution images and videos of your specimens.
- 20x to 200x powers, great for low-power observation of 3D specimens
- Professional, adjustable height stand included for hands-free operation
- 4 foot USB 2.0 cable for easy maneuverability when viewing large surfaces
- Intuitive software with measuring features
- Windows and Mac compatible
The Celestron Handheld Digital Microscope Pro is an easy to use, low-power microscope with a 5.0 MP sensor for capturing photos and video of your discoveries. At low magnification, the Handheld Digital Microscope Pro is ideal for viewing stamps, coins, bugs, plants, rocks, skin, gems, circuit boards, and more. You can even view traditional slides at high magnification.
Create high-resolution images, VGA quality video, and time-lapse animations using the included Micro Capture Pro software. You can also take accurate measurements and make notes on your specimens using this intuitive software. An adjustable LED illuminator ensures your specimens are clear and bright, while the fully adjustable, professional stand allows for steady shots even at high magnification.
Intended for the discriminating hobbyist or professional, the Handheld Digital Microscope Pro is well suited for jobs such as entomology, industrial quality control, component inspection, medical investigation, scientific research, and forensics documentation—just to name a few.
Note: Final magnification determined by monitor size.
Documents
Especificaciones
- Magnification:20x to 200x
- Model:44308
- Description:Handheld Digital Microscope Pro