Sterile SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
76097-918
- Esterilidad:Sterile
- Anchura:22.9 cm (9")
- Longitud:27.9 cm (11")
- Matriz:PP
- Referencia:76097-918
- Empaquetado:50 Wipes/Pack
Sterile SatPax® 550 wipers are composed of exceptionally clean, 100% meltblown polypropylene.
- Designed for use in ISO Class 5 and higher cleanroom environments
- Very low extractables and ions
- Very low particle and fiber counts
- Reduces alcohol usage and preparation/handling costs
- Reduces VOC emissions
- Increases cleaning efficiency
- Increases cleaning protocol consistency
- Resealable solvent-resistant packaging
Sterile SatPax® 550 wipers are composed of exceptionally clean, 100% meltblown polypropylene, pre-wetted with 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper. Each package is gamma irradiated sterile to a 10⁻⁶ Sterility Assurance Level, making it ideal for cleaning critical sterile processing environments.
: Gamma irradiated and sterile validated to a 10⁻⁶ Sterility Assurance Level per AAMI guidelines. ISO Class 5 – 8, Cleanroom Class 100 – 100,000, EU Grade A/B/C/D. EC Regulation 1935 (2004)- Food contact compliant. FDA 21 CFR-Food contact compliant.