Orders LinkContactUsLinkComponent
DR-MED® Laparoscopic Training Instruments
DR-MED® Laparoscopic Training Instruments
DR-MED® Laparoscopic Training Instruments
Certificates
DR-MED® Laparoscopic Training Instruments
DR-MED® Laparoscopic Training Instruments
4 Options Available Below
Product Details & Documents
Reasonably priced practice tools.
  • The same tools you would use in surgery
  • 360 degree rotation
  • 19" length
Learn skills using tools with the same features and grips as the surgical instruments you will be using. Securely hold and manipulate tissues with the grasper and perform fine manipulations with the forceps. Practice removing tissue with the scissors, and closing internally with the needle driver.
Caution: For Training Purposes Only. Not for human or animal use.
Product Family Options
Información del producto
  • Availability
    Price
  • 470231-224
    Laparoscopic Scissors
    Laparoscopic Scissors
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,728.37
    $3,728.37
    Each (1UN)
       $3,728.37
    Supplier Number: DR786924
  • 470231-226
    Laparoscopic Needle Holder
    Laparoscopic Needle Holder
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,450.60
    $4,450.60
    Each (1UN)
       $4,450.60
    Supplier Number: DR786920
  • 470231-228
    Laparoscopic forceps
    Laparoscopic forceps
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,861.63
    $3,861.63
    Each (1UN)
       $3,861.63
    Supplier Number: DR786922
  • 470231-230
    Laparoscopic Grasper
    Laparoscopic Grasper
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,728.37
    $3,728.37
    Each (1UN)
       $3,728.37
    Supplier Number: DR786923