- Resolución:T: 0.01 °C
- Interfaz:WiFi
- Descripción:1 stainless steel 316 sensor with 6" cable and platinum RTD sensor
- Pilas:4 AAA Alkaline battery
- Peso:113 g
- Ancho × Fondo × Alto:108×70×19 mm (4.25×2.75×0.75")
- Referencia:76047-268
- Exactitud:T: ±0.25 °C
- Rango:T: –200...+105 °C (–328...+221°F)
Covering cryo needs for sensitive, irreplaceable bioligicals.
- Monitor temperature readings locally and remotely by connecting to existing Wi-Fi
- Ideal for LN₂ storage vessels, 2 probe version permits monitoring the LN₂ dewar and ambient air temperature simultaneously
- Alerts: temperature alarm, connectivity interruption, low battery
- Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
- Continuous data transmission and monitoring using SSL encrypted SECURE connection
- Receive Mobile Push, Email, or Text notifications
- Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
- Multiple point temperature calibration traceable to NIST, with calibration points of –196°C, –80°C, 0°C, and 100°C
Platinum sensor, stainless-steel probe has fast-response platinum RTD sensor, stainless-steel 316 probe displays the slightest temperature change instantly. Place probe inside dewar-readings are updated three times a second. Probe has a Ø of 0.12", stem length of 6" and a cable length of 10'.
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go. Audible and visual alarms helps those present in the lab identify any out-of-range conditions. High-accuracy thermometer, high-impact, chemical-resistant ABS plastic case. Ideal for monitoring long term sensitive biologicals, stem cell and more., that are irreplaceable. Works with any liquid nitrogen/cryogenic storage, dewar.
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on website (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).