Especificaciones
- Resolución:T: 0.01 °C
- Descripción:1 stainless steel 316 sensor with 6" cable and platinum RTD sensor
- Peso:113 g
- Ancho × Fondo × Alto:108×70×19 mm (4.25×2.75×0.75")
- Referencia:76047-254
- Exactitud:T: ±0.1 °C
- Rango:T: –90...+105 °C (–130...+221°F)
Especificaciones
Acerca de este artículo
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go.
- Monitor temperature readings locally and remotely by connecting to existing Wi-Fi
- Ideal for ULT freezers, two probe version permits monitoring the temperature of a ULT freezer and ambient air temperature simultaneously
- Alerts: temperature alarm, connectivity interruption, low battery
- Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
- Continuous data transmission and monitoring using SSL encrypted SECURE connection
- Receive Mobile Push, Email, or Text notifications
- Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
- Multiple point temperature calibration traceable to NIST, with calibration points of −80, −40, 0, and 100 °C
Stainless-steel probe: Fast-response stainless-steel 316 probe displays the slightest temperature change instantly. Place probe inside ULT freezer-readings are updated six times a minute. Probe has a Ø of 0.12", stem length of 6", dimension of 5.875" and a cable length of 9'.
Audible, high-accuracy thermometer and visual alarms helps those present in the lab identify any out-of-range conditions. Designed for Ultra-Low (ULT) freezers-monitor temperatures below −80 °C ensuring critical samples are not compromised. Ideal for monitoring ultra-low temperature freezers and include a FEP cable. High-impact and chemical-resistant ABS plastic case.
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on website (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).