ContactUsLinkComponent
Three-Neck Reaction Vessel Lids, Chemglass
  80071-568
Small Business Enterprise
undefined
Three-Neck Reaction Vessel Lids, Chemglass
  80071-568
 :  CG-1940-03
Small Business Enterprise
  • Descripción:
    Three-neck reaction vessel lid, 150 mm flange
  • Referencia:
    80071-568
  • Side ST Joint Size:
    24/40
  • Center Joint:
    24/40
  • Nº de proveedor:
    CG-1940-03
These three neck reaction vessel lids have a flat ground surface allowing it to be greased or used with an o-ring.
 : Three neck lids are available with joint sizes and configurations other than those listed.