Funnels, Separatory, Squibb, PTFE Stopcock, With Sealed Drip Tip, Chemglass
80071-150
Small Business Enterprise
- Capacidad:250 ml
- Descripción:Funnel, PTFE Stopcock, With Sealed Drip Tip
- Referencia:80071-150
- ST Joint Size:24/40
- Tamaño del tapón:4 mm
- Nº de proveedor:CG-1750-03
Squibb-style separatory funnel with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip.
: Supplied complete with a flask length PTFE stopper.