- Ø:30 mm
- Tamaño de poro:0.70 µm
- Color:Amber
- Referencia:14235-016
- Membrana:Glass MicroFiber
- Nº de proveedor:40725-GM
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
- For large particulate removal
- Increased sample throughput
- Low extraction neutral borosilicate glass
- Able to filter solutions at temperatures up to 110 °C
- For use with viscous or particle-laden samples
Available in sizes and membrane types to meet any application need, Thermo Scientific™ Titan3™ GMF patented Syringe Filters provide cleaner sample extracts by removing interfering materials and fine particles.
Glass microfiber filter applications are clarification and pre-filtering of suspensions GMF filters are available in a range of porosities.
Low extractable membranes and housings ensure that no filter-borne contaminants are added to your sample. Reinforcing ring provides greater strength to the housing to prevent leakage and bursting (maximum pressure for 30 mm devices 15 bar / 210 psi). Housings are manufactured from solvent-resistant, low-extractable polypropylene resins specifically selected for wide compatibility with common HPLC sample matrices.
Color coding makes it easy to select the correct membrane and pore size. Packed in re-usable rigid transparent color-coded containers. Syringe filters can be sterilized by autoclave at 121 °C for 15 minutes. Enhanced Luer lock inlet prevents leakage; outlet fitting is a standard size male Luer-slip fitting for ease of filtrate collection.