Kuderna-Danish Concentrator Tubes, Graduated, Chemglass
80068-040
Small Business Enterprise
- Capacidad:25 ml
- Descripción:Kuderna-Danish Concentrator Tube, Graduated, without hooks
- Altura:170 mm
- Referencia:80068-040
- Intervalo de graduación:0.1 (0 - 1 ml)/1.0 (2 - 25 ml)
- [ST] unión No.:19/22
- Nº de proveedor:CG-1316-12
These graduated concentrator tubes are mainly designed for use with Kuderna-Danish evaporative set-ups.
- Can be stoppered for storage
- Available with and without hooks