Adapter, Distillation, Chemglass
80065-816
Small Business Enterprise
- Descripción:Distilling Adapter
- Referencia:80065-816
- ST Joint Size:24/40
- Nº de proveedor:CG-1022-01
These adapters for distillation systems are designed with standard taper outer joint at the top and inner joints on the bottom and sidearm.
- Inner joints are at a 75° angle to one another
- All the joints are in same size