VWR® Erlenmeyer Flask with GL 45 Screw Cap
Proveedor: VWR International
Certificados
Acerca de este artículo
Flask is suitable for mixing liquids.
- Very good chemical resistance
- Scale graduations
- Autoclavalable
Typical uses include substance storage, media preparation, and cultivation. Made from high quality Borosilicate glass 3.3.
Tolerance: 2 mm
Información de pedido: Flasks are supplied with polypropylene GL45 thread caps.
Información relacionada
Certificados
Buscar por número de loteCoQ VWR Erlenmeyer Flask with GL 45 Screw Caps.pdf
Declaración de conformidad
Frequently Bought Together