AEA Sporulation Medium Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95020-760
Proveedor: HIMEDIA LABORATORIES LLC
Número CAS:
Especificaciones
- Descripción:AEA Sporulation Medium Base, Modified
- Referencia:95020-760
- Nº de proveedor:M1236-500G
Especificaciones
Acerca de este artículo
For the early sporulation of Clostridium perfringens from foods.
- Microbiological culture media
- Packaged in 500 g poly bottles