ContactUsLinkComponent
EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Catalog # 10827-860
Supplier:  VWR International
undefined
EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Catalog # 10827-860
Supplier:  VWR International
Specifications
  • Descripción:
    EZwaste® System, 83 mm Cap, 6x 1/8" OD Tubing, 1x 1/4 or 3/8" ID Hose Barb and Filter
  • Referencia:
    10827-860
Specifications
About this item
This cap system allows for the safe and effective collection of waste in the laboratory
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb option in 1/4" or 3/8"
The EZwaste® Solvent Waste System is designed to be an affordable solution forcontaining liquid chromatography waste. Each system is manufactured to safely collect waste from one or more HPLC systems while the carbon exhaust filters improve air quality by capturing potentially harmful vapors emitted from solvents. Each cap configuration includes an 83 mm VersaCap®, open top for waste collection, 6 ports with a variety of O.D. configurations for waste tubes, one port for a carbon exhaust filter, one carbon exhaust filter, and plugs for unused ports. Tube fittings secure waste tubes to the carboy, providing the closed system necessary to prevent spills (plugs included for unused ports). A 90° adapter is included for maximum storage flexibility.
The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.