Orders LinkContactUsLinkComponent
Neodymium Disk Magnet
Neodymium Disk Magnet
Neodymium Disk Magnet
Neodymium Disk Magnet
Neodymium Disk Magnet
A powerful magnet for a variety of experiments.
  • Very high resistance to demagnetization
  • High energy for size
  • Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. All of the magnets have a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).
Product Family Options
  • Description
  • Neodymium Disk Magnet, 1" Diameter, 0.74 " Thick
    Each (1UN)
    Each (1UN) - $381.63
    $381.63
    Each (1UN)
       $381.63
     : ND030N-35
  • Neodymium Disk Magnet, 0.5" Diameter, 0.25 " Thick
    Paquete de 3
    Paquete de 3 - $125.26
    $125.26
    Paquete de 3
       $125.26
     : ND143N-35BXX3
  • Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick
     : -