EZFlow® Syringe Filter, Glass Fiber, Foxx Life Sciences
76018-900
- Ø:25 mm
- Tamaño de poro:0.22 µm
- Esterilidad:Non sterile
- Referencia:76018-900
Glass Fiber syringe filters have super low extractables and fast flow rates.
- Chemically inert membrane
- Super low extractables
- Fast flow rates
- Resistant to almost all solvents
EZFlow® 13, 25, and 33 mm syringe filters are used for prefiltration, clarification, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials, in 0.2 or 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.
They are chemically inert and resistant to almost all solvents.
: All materials comply with USP Class VI Standards and FDA compliances.