Microflex® N85 Nitrile Exam Gloves, Ansell
Microflex® N85 is a nitrile glove with a soft and stretchy feel.
- Fully textured, it protects while providing a reliable grip
- Provides a reliable grip
Recommended uses include all food processing and applications that have food contact, assembly of parts, equipment repair and maintenance, general purpose auto aftermarket, maintaining lab furniture and equipment, sample taking and processing, and inspection, selecting, and checking parts.
- Catalog No:
- 10119-318
- 10119-320
- 10119-316
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Espesor del puño:
- 2.4 mils
- 2.4 mils
- 2.4 mils
- Espesor del dedo:
- 4.3 mils
- 4.3 mils
- 4.3 mils
- Espesor de la palma:
- 3.1 mils
- 3.1 mils
- 3.1 mils
- Alargamiento de rotura:
- 500% (before aging), 400% (after aging)
- 500% (before aging), 400% (after aging)
- 500% (before aging), 400% (after aging)
Frequently Bought Together