Orders LinkContactUsLinkComponent
Graduated Reagent Bottle
Graduated Reagent Bottle
Certificates
Graduated Reagent Bottle
Graduated Reagent Bottle
10 Options Available Below
Product Details & Documents
Mess-free pouring.
  • Screw cap
  • Borosilicate glass
  • Poring ring
  • Both cap and bottle are autoclavable
These economy graduated reagent bottles are the perfect addition to any laboratory. Made from borosilicate glass, these bottles have a convienent screw cap and a drip-free pouring ring to help maintain a clean work environment.
Product Family Options
Información del producto
  • Capacity
    Color
    Availability
    Price
  • 100 ml
    Clear
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $255.84
    $255.84
    Each (1UN)
       $255.84
    Supplier Number: CH0164A
  • 250 ml
    Clear
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $340.06
    $340.06
    Each (1UN)
       $340.06
    Supplier Number: CH0164B
  • 500 ml
    Clear
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $408.84
    $408.84
    Each (1UN)
       $408.84
    Supplier Number: CH0164C
  • 1000 ml
    Clear
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $874.66
    $874.66
    Each (1UN)
       $874.66
    Supplier Number: CH0164D
  • 2000 ml
    Clear
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,373.55
    $1,373.55
    Each (1UN)
       $1,373.55
    Supplier Number: CH0164E
  • 100 ml
    Amber
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $371.49
    $371.49
    Each (1UN)
       $371.49
    Supplier Number: CH0165A
  • 250 ml
    Amber
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $506.35
    $506.35
    Each (1UN)
       $506.35
    Supplier Number: CH0165B
  • 500 ml
    Amber
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $611.88
    $611.88
    Each (1UN)
       $611.88
    Supplier Number: CH0165C
  • 1000 ml
    Amber
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,132.10
    $1,132.10
    Each (1UN)
       $1,132.10
    Supplier Number: CH0165D
  • 2000 ml
    Amber
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,745.05
    $1,745.05
    Each (1UN)
       $1,745.05
    Supplier Number: CH0165E