Orders LinkContactUsLinkComponent
Graduated Cylinders
Graduated Cylinders
Graduated Cylinders
Certificates
Graduated Cylinders
Graduated Cylinders
6 Options Available Below
Product Details & Documents
Class A Cylinders.
  • Blue graduations
  • Hexagonal base
  • Plastic cylinder guard
These Class A Graduated Cylinders are the perfect laboratory companion. Made from high quality borosilicate glass with a hexagonal base. Supplied complete with protective cylinder guard. Cylinders comply with ASTM E1272 standard.
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Tolerance
    Availability
    Price
  • 10 ml
    0.2 ml
    ±0.10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $147.65
    $147.65
    Each (1UN)
       $147.65
    Supplier Number: CH0341ABL
  • 25 ml
    0.5 ml
    ±0.17 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $275.57
    $275.57
    Each (1UN)
       $275.57
    Supplier Number: CH0341BBL
  • 50 ml
    1.0 ml
    ±0.25 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $308.60
    $308.60
    Each (1UN)
       $308.60
    Supplier Number: CH0341CBL
  • 100 ml
    1.0 ml
    ±0.50 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $385.88
    $385.88
    Each (1UN)
       $385.88
    Supplier Number: CH0341DBL
  • 500 ml
    5.0 ml
    ±2.00 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $746.74
    $746.74
    Each (1UN)
       $746.74
    Supplier Number: CH0341FBL
  • 1000 ml
    10.0 ml
    ±3.00 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,388.49
    $1,388.49
    Each (1UN)
       $1,388.49
    Supplier Number: CH0341GBL