Orders LinkContactUsLinkComponent
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller
Supplier:   MTC BIO, INC.
ProPette™ Electronic Pipette Controller
Supplier:  MTC BIO, INC.
Certificates
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller
5 Options Available Below
Product Details & Documents
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
  • QuickStand™ prevents pipette contamination
  • Variable speed control
  • Cordless and rechargable
  • Provides 8 hours of continuous operation
  • Fits serological pipettes from 1 to 100 ml
  • Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.
Product Family Options
Electronic Pipette Controller
Información del producto
  • Availability
    Price
  • 470228-478
    Pipette Controller, with US Plug
    Pipette Controller, with US Plug
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $8,076.66
    $8,076.66
    Each (1UN)
       $8,076.66
    Supplier Number: P6080
Replacement Parts
Información del producto
  • Availability
    Price
  • 470228-482
    Lithium Battery
    Lithium Battery
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $794.72
    $794.72
    Each (1UN)
       $794.72
    Supplier Number: P6080-BA
  • 470228-484
    Filters (0.45 µm)
    Filters (0.45 µm)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 5
    Paquete de 5 - $874.14
    $874.14
    Paquete de 5
       $874.14
    Supplier Number: P6080-FI
  • 470228-486
    Silicone Nosepiece
    Silicone Nosepiece
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $690.25
    $690.25
    Each (1UN)
       $690.25
    Supplier Number: P6080-NA
  • Charger
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $794.72
    $794.72
    Each (1UN)
       $794.72
    Supplier Number: P6080-RC