Avantor® Prevail C18-Select, HPLC Columns
75798-960
: Avantor
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:5 µm
- Longitud:150 mm
- Environmentally Preferable:
- Referencia:75798-960
- Packing Material:Silica
- Phase:C18-Select
- Nº de proveedor:99300
Avantor® Hichrom Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases.
- Stable from highly organic to highly aqueous
- Specialty phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
- USP L1
Same benefits as Avantor® Hichrom Prevail™ C18, but slightly more retentive.
All phases, apart from Avantor® Hichrom Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
: Use guard cartridges with holder (80101/N) and coupler (HI-081).