ContactUsLinkComponent
Vydac 201TP C18, HPLC Columns
  75798-274
undefined
Vydac 201TP C18, HPLC Columns
  75798-274
  • Formato de columna:
    Analytical
  • Ø int.:
    4.60 mm
  • Tamaño de partícula:
    5 µm
  • Tamaño de poro:
    300 Å
  • Longitud:
    250 mm
  • Environmentally Preferable:
  • Extremo tapado:
    No
  • Código USP:
    L1
  • Carga de carbono:
    8%
  • Column Hardware:
    Acero inoxidable
  • Referencia:
    75798-274
  • Packing Material:
    Silica
  • Phase:
    C18
  • Nº de proveedor:
    201TP54
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
Non-endcapped C18 phase. Developed for the separation of PAHs.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
Guard holders and accessories