SHOWA 540 Polyurethane Palm Coated, A2 Cut Resistant Glove, White, Showa
94002-646
- Color del guante:White
- Tamaño del guante:M
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Desechable / Reutilizable:Reutilizable
- Latex/No contiene Latex:No contiene Latex
- Con polvo/sin polvo:Sin polvo
- Longitud:27.9 cm (11")
- Tipo de puño:Puño elástico
- Referencia:94002-646
- Nº de proveedor:540-M
A2 cut resistant gloves offer extreme dexterity, grip, and comfort.
- Ergonomic design
- Excellent grip in oil
- A2 cut resistance
A2 Polyurethane palm coated gloves are excellent where dexterity and comfort are needed for maintenance, assembly, and general-purpose work.