Vydac 218TP C18, HPLC Columns
Proveedor: Avantor
Certificados
Acerca de este artículo
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
- USP L1
First generation polymeric C18 phase with unique selectivity. Recommended for small polypeptides 4-5K MW, enzymatic digest fragments, natural and synthetic peptides, multi-ring compounds.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.
Guard Cartridge Holders
Información del producto
Frequently Bought Together