Cometron 7X50
Ideal binocular for comet watching, stargazing, and viewing craters on the Moon.
- BK7 Prism Glass
- 7X magnification
- 50 mm objective lens diameter
- Wide neck strap and carrying case included
Wide field of view reveals a larger portion of the night sky, allowing you to view more of the comet's impressive tail. Large 50mm objective lenses have tremendous light-gathering ability, ideal for astronomical use. Multi-coated optics dramatically increase light transmission for brighter images with increased contrast and resolution. Large exit pupil provides maximum light during nighttime and low-light viewing sessions. Strong and durable alumnium housing.
- Magnification:7×
- Description:Cometron 7X50