Process2Wipe® VAI® IPA70 Saturated Wipes
Process2Wipe, 30.5×30.5 cm (12×12"), Sterile
Supplier: VELTEK ASSOCIATES, INC. MX
Número de proveedor #: VEL12-12X12-S-3023
Jump to:
Product Details & Documents
Process2Wipes are saturated with 70% USP isopropyl alcohol and 30% USP water for injection.
- 100% continuous polyester fiber is clean and absorbent
- ISO 4 (Class 10) cleanliness
- Validated for sterility and shelf life
These wipes are used for aseptic and non-aseptic wipe downs of filling and packaging machinery, stainless steel surfaces, Lexan, polycarbonate, glass, and any critical surfaces that require cleaning.
Packaging: Delivered with lot specific documentation. Resealable storage packaging.
Documents
Certificates
Buscar por número de loteVWR GHS Compliance Letter (US)
Detalles adicionales del producto
Especificaciones
- Sterility:Sterile
- Description:Process2Wipe
- W×L:30.5×30.5 cm (12×12")